Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATTTGGAAGTCACTGGGTCAGA[G/T]CCTGTATCACATCCTTTACCAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602166 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3B2 - adaptor related protein complex 3 beta 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278511.1 | 3495 | Missense Mutation | GAT,GCT | D,A 1047 | NP_001265440.1 | |
NM_001278512.1 | 3495 | Missense Mutation | GAT,GCT | D,A 1098 | NP_001265441.1 | |
NM_004644.4 | 3495 | Missense Mutation | GAT,GCT | D,A 1079 | NP_004635.2 | |
XM_017022640.1 | 3495 | Missense Mutation | GAT,GCT | D,A 1055 | XP_016878129.1 | |
XM_017022641.1 | 3495 | Intron | XP_016878130.1 |
CPEB1-AS1 - CPEB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |