Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGGTTCACATGACCGCACAACA[C/T]TGGCACGTCTTTTGCTTAGGGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL27RA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL27RA - interleukin 27 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004843.3 | 2004 | Intron | NP_004834.1 |
PALM3 - paralemmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145028.1 | 2004 | Silent Mutation | CAA,CAG | Q,Q 668 | NP_001138500.1 | |
XM_005259891.4 | 2004 | Silent Mutation | CAA,CAG | Q,Q 683 | XP_005259948.1 | |
XM_011527965.2 | 2004 | Silent Mutation | CAA,CAG | Q,Q 656 | XP_011526267.1 |