Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACGTTCTCTTGATTCACATCC[A/G]TAGGAGAGTCTAGGGTAAAGCGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAC3 - tachykinin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178054.1 | 481 | Missense Mutation | ACG,ATG | T,M 83 | NP_001171525.1 | |
NM_013251.3 | 481 | Missense Mutation | ACG,ATG | T,M 101 | NP_037383.1 |
ZBTB39 - zinc finger and BTB domain containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |