Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTTGTTCAGCTTTTCCAGGGCT[G/T]CGTAGAAGTTTTCATAAGGGATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929710 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929710 - uncharacterized LOC101929710 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCSK1 - proprotein convertase subtilisin/kexin type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000439.4 | 2344 | Missense Mutation | GAA,GCA | E,A 702 | NP_000430.3 | |
NM_001177875.1 | 2344 | Missense Mutation | GAA,GCA | E,A 655 | NP_001171346.1 |