Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAACAAAGACCTATTTTATCTCA[A/G]TGTCACACAGACAAGTGGCTTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRISP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRISP1 - cysteine rich secretory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131.2 | 765 | Missense Mutation | ACT,ATT | T,I 246 | NP_001122.2 | |
NM_001205220.1 | 765 | Missense Mutation | ACT,ATT | T,I 246 | NP_001192149.1 | |
NM_170609.1 | 765 | UTR 3 | NP_733758.1 | |||
XM_011514315.2 | 765 | Missense Mutation | ACT,ATT | T,I 144 | XP_011512617.1 | |
XM_017010320.1 | 765 | Missense Mutation | ACT,ATT | T,I 144 | XP_016865809.1 | |
XM_017010321.1 | 765 | Missense Mutation | ACT,ATT | T,I 144 | XP_016865810.1 |