Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAATTCCTGTTTAACAATACAG[A/G]GTCTCTTGCAACGTGTGAGCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSRB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSRB2 - methionine sulfoxide reductase B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012228.3 | 230 | Missense Mutation | GAG,GGG | E,G 40 | NP_036360.3 | |
XM_011519426.2 | 230 | Missense Mutation | GAG,GGG | E,G 40 | XP_011517728.1 |