Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACATAAAACCCTCACAAACCTCA[A/G]GAAACCAGGGCTAAAAGAACAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP4S1 - adaptor related protein complex 4 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HECTD1 - HECT domain E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015382.3 | 7441 | Silent Mutation | CTG,TTG | L,L 2521 | NP_056197.3 | |
XM_005267502.2 | 7441 | Silent Mutation | CTG,TTG | L,L 2521 | XP_005267559.2 | |
XM_011536619.2 | 7441 | Silent Mutation | CTG,TTG | L,L 2519 | XP_011534921.1 | |
XM_011536621.2 | 7441 | Silent Mutation | CTG,TTG | L,L 2242 | XP_011534923.1 | |
XM_011536622.1 | 7441 | Silent Mutation | CTG,TTG | L,L 1971 | XP_011534924.1 | |
XM_017021148.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2474 | XP_016876637.1 | |
XM_017021149.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2472 | XP_016876638.1 | |
XM_017021150.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2460 | XP_016876639.1 | |
XM_017021151.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2242 | XP_016876640.1 | |
XM_017021152.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2240 | XP_016876641.1 | |
XM_017021153.1 | 7441 | Silent Mutation | CTG,TTG | L,L 2240 | XP_016876642.1 |