Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCGGGCAGCCCGGCGCTGGAA[C/G]TGCACCAAGGGCTGGGAGCGGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM171A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM171A2 - family with sequence similarity 171 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198475.2 | 185 | Missense Mutation | CAC,CAG | H,Q 155 | NP_940877.2 | |
XM_017024490.1 | 185 | UTR 5 | XP_016879979.1 |
GRN - granulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |