Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGTGCCCTGCGTGAAGAGCATC[A/G]TGAGGCAGGTGAGTGCCACCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300089 MIM: 300214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDH3G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDH3G - isocitrate dehydrogenase 3 (NAD(+)) gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLXNB3 - plexin B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRPK3 - SRSF protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170760.1 | 660 | Missense Mutation | ATG,GTG | M,V 192 | NP_001164231.1 | |
NM_001170761.1 | 660 | Missense Mutation | ATG,GTG | M,V 192 | NP_001164232.1 | |
NM_014370.3 | 660 | Missense Mutation | ATG,GTG | M,V 192 | NP_055185.2 |