Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGGAAGGCTCAGGCTGGACAC[C/G]TATTGTATGAGGTGGGCCTCAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LFNG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LFNG - LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040167.1 | 162 | Intron | NP_001035257.1 | |||
NM_001040168.1 | 162 | Intron | NP_001035258.1 | |||
NM_001166355.1 | 162 | Silent Mutation | ACC,ACG | T,T 12 | NP_001159827.1 | |
NM_002304.2 | 162 | Intron | NP_002295.1 |