Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCAAACGTGTAACTGGTTCATC[A/G]GTGTCCAGATGATTAAGATCCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR89 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR89 - WD repeat domain 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008726.2 | 1515 | Silent Mutation | ACC,ACT | T,T 251 | NP_001008726.1 | |
NM_001258272.1 | 1515 | Silent Mutation | ACC,ACT | T,T 251 | NP_001245201.1 | |
NM_080666.3 | 1515 | Silent Mutation | ACC,ACT | T,T 251 | NP_542397.1 | |
XM_011536383.2 | 1515 | Silent Mutation | ACC,ACT | T,T 289 | XP_011534685.1 | |
XM_011536385.2 | 1515 | Silent Mutation | ACC,ACT | T,T 251 | XP_011534687.1 |