Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCACGTTGTGTCACATTCTAT[C/T]GAAACCTGATTTTCACGTACTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927179 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927179 - uncharacterized LOC101927179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFXL1 - nuclear transcription factor, X-box binding like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278623.1 | 2557 | Silent Mutation | TCA,TCG | S,S 820 | NP_001265552.1 | |
NM_001278624.1 | 2557 | Silent Mutation | TCA,TCG | S,S 820 | NP_001265553.1 | |
NM_152995.5 | 2557 | Silent Mutation | TCA,TCG | S,S 820 | NP_694540.3 |