Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGATGCTATAAGGTAAATGTTA[C/T]GCAAAGATGACAACTGACCAATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ORC2 - origin recognition complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006190.4 | 1580 | Missense Mutation | CAT,CGT | H,R 416 | NP_006181.1 | |
XM_006712555.3 | 1580 | Missense Mutation | CAT,CGT | H,R 416 | XP_006712618.1 | |
XM_011511252.2 | 1580 | Missense Mutation | CAT,CGT | H,R 436 | XP_011509554.1 | |
XM_011511253.2 | 1580 | Missense Mutation | CAT,CGT | H,R 436 | XP_011509555.1 |