Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCCACCTGTTCGTAGATGTCA[A/G]TGGCCTTCTGATACTGCTCCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615620 MIM: 603215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KPTN - kaptin, actin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPA - NSF attachment protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003827.3 | 537 | Missense Mutation | ACT,ATT | T,I 182 | NP_003818.2 | |
XM_011527436.1 | 537 | Missense Mutation | ACT,ATT | T,I 132 | XP_011525738.1 | |
XM_011527437.1 | 537 | Missense Mutation | ACT,ATT | T,I 78 | XP_011525739.1 |
NAPA-AS1 - NAPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |