Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTGGTCTTGAAGAGCAGCTTAG[C/T]GAAAGAAACCCTCTCTGAGTTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP9 - aquaporin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320635.1 | 438 | Missense Mutation | GCG,GTG | A,V 23 | NP_001307564.1 | |
NM_001320636.1 | 438 | Intron | NP_001307565.1 | |||
NM_020980.4 | 438 | Missense Mutation | GCG,GTG | A,V 23 | NP_066190.2 |
LOC101928635 - uncharacterized LOC101928635 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011522293.2 | 438 | Intron | XP_011520595.1 |