Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCTGGCTGTGTGTCCCTGCAGG[C/T]GGAGCTAAATGAGGTCCTTGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf57 - chromosome 19 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CC2D1A - coiled-coil and C2 domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017721.4 | 613 | Missense Mutation | GCG,GTG | A,V 105 | NP_060191.3 | |
XM_005259972.2 | 613 | Missense Mutation | GCG,GTG | A,V 105 | XP_005260029.1 | |
XM_005259973.2 | 613 | Missense Mutation | GCG,GTG | A,V 105 | XP_005260030.1 | |
XM_005259974.2 | 613 | Missense Mutation | GCG,GTG | A,V 105 | XP_005260031.1 | |
XM_005259975.2 | 613 | Missense Mutation | GCG,GTG | A,V 105 | XP_005260032.1 |