Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATTAAAAGGTATTACCTGTTGG[C/T]TGAAGTCCAGAGTGTCCTGGGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 142857 | ||||||||||||||||||||
Literature Links: |
HLA-DRB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-DRB1 - major histocompatibility complex, class II, DR beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243965.1 | 874 | Silent Mutation | CAA,CAG | Q,Q 260 | NP_001230894.1 | |
NM_002124.3 | 874 | Silent Mutation | CAA,CAG | Q,Q 260 | NP_002115.2 |