Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATCATAAATAGAATTTACCTTC[C/T]TGTATCCCACAGCTTAACAATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR12 - WD repeat domain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018256.3 | 1744 | Missense Mutation | AGA,GGA | R,G 373 | NP_060726.3 | |
XM_011511469.2 | 1744 | Missense Mutation | AGA,GGA | R,G 235 | XP_011509771.1 |