Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCTCTCCCCTTCAGGTTTTCA[C/T]GGTGCTCCTCATGCCTCCGCCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC149 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC149 - coiled-coil domain containing 149 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248210.3 | 1695 | Missense Mutation | XP_005248267.1 | |||
XM_011513906.2 | 1695 | Missense Mutation | XP_011512208.1 | |||
XM_011513907.2 | 1695 | Missense Mutation | XP_011512209.1 | |||
XM_011513908.1 | 1695 | Missense Mutation | XP_011512210.1 | |||
XM_017008827.1 | 1695 | Missense Mutation | XP_016864316.1 | |||
XM_017008828.1 | 1695 | Missense Mutation | XP_016864317.1 |
SOD3 - superoxide dismutase 3, extracellular | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |