Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTCTTCATACATAGCTTTCTTT[C/T]ACTTTCTCATCCTGTAGAAAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600046 MIM: 608872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA1 - ATP binding cassette subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005502.3 | 7087 | Silent Mutation | GTA,GTG | V,V 2256 | NP_005493.2 | |
XM_005251773.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2258 | XP_005251830.1 | |
XM_005251776.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2198 | XP_005251833.1 | |
XM_011518339.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_011516641.1 | |
XM_011518340.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_011516642.1 | |
XM_011518341.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2281 | XP_011516643.1 | |
XM_011518342.2 | 7087 | Silent Mutation | GTA,GTG | V,V 2137 | XP_011516644.1 | |
XM_011518344.2 | 7087 | Intron | XP_011516646.1 | |||
XM_017014378.1 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_016869867.1 | |
XM_017014379.1 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_016869868.1 | |
XM_017014380.1 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_016869869.1 | |
XM_017014381.1 | 7087 | Silent Mutation | GTA,GTG | V,V 2283 | XP_016869870.1 | |
XM_017014382.1 | 7087 | Silent Mutation | GTA,GTG | V,V 2237 | XP_016869871.1 |
NIPSNAP3B - nipsnap homolog 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |