Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGAGGTGGGGGCCCACCAGGAT[A/G]AGCAAGCTGCCCAGGGAGCTGACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607048 | ||||||||||||||||||||
Literature Links: |
STARD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STARD3 - StAR related lipid transfer domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165937.1 | 206 | Missense Mutation | ATA,ATG | I,M 1 | NP_001159409.1 | |
NM_001165938.1 | 206 | Missense Mutation | ATA,ATG | I,M 1 | NP_001159410.1 | |
NM_006804.3 | 206 | Missense Mutation | ATA,ATG | I,M 1 | NP_006795.3 | |
XM_006721646.2 | 206 | Missense Mutation | ATA,ATG | I,M 1 | XP_006721709.1 | |
XM_011524210.1 | 206 | Missense Mutation | ATA,ATG | I,M 1 | XP_011522512.1 | |
XM_011524211.2 | 206 | Missense Mutation | ATA,ATG | I,M 1 | XP_011522513.1 | |
XM_011524213.1 | 206 | Missense Mutation | ATA,ATG | I,M 1 | XP_011522515.1 | |
XM_017024041.1 | 206 | Missense Mutation | ATA,ATG | I,M 1 | XP_016879530.1 |