Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTTGGATGGATTCATTAAAAA[A/G]TTTGTTCTCTGTCCTGAATGTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601710 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF5 - eukaryotic translation initiation factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001969.4 | 818 | Silent Mutation | AAA,AAG | K,K 95 | NP_001960.2 | |
NM_183004.4 | 818 | Silent Mutation | AAA,AAG | K,K 95 | NP_892116.2 |
LOC105370687 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA28 - small nucleolar RNA, H/ACA box 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |