Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF423 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF423 - zinc finger protein 423 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271620.2 | 5796 | UTR 3 | NP_001258549.1 | |||
NM_015069.4 | 5796 | UTR 3 | NP_055884.2 | |||
XM_005255856.4 | 5796 | Intron | XP_005255913.1 | |||
XM_005255857.4 | 5796 | Intron | XP_005255914.1 | |||
XM_006721171.3 | 5796 | UTR 3 | XP_006721234.1 | |||
XM_017023076.1 | 5796 | UTR 3 | XP_016878565.1 | |||
XM_017023077.1 | 5796 | Intron | XP_016878566.1 | |||
XM_017023078.1 | 5796 | Intron | XP_016878567.1 |