Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601507 MIM: 609608 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3S1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AP3S1 - adaptor related protein complex 3 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002924.2 | Intron | NP_001002924.2 | ||||
NM_001284.3 | Intron | NP_001275.1 | ||||
NM_001318090.1 | Intron | NP_001305019.1 | ||||
NM_001318091.1 | Intron | NP_001305020.1 | ||||
NM_001318093.1 | Intron | NP_001305022.1 | ||||
NM_001318094.1 | Intron | NP_001305023.1 | ||||
XM_011543136.2 | Intron | XP_011541438.2 | ||||
XM_017009024.1 | Intron | XP_016864513.1 | ||||
XM_017009025.1 | Intron | XP_016864514.1 |
ATG12 - autophagy related 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |