Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300617 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRCC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BRCC3 - BRCA1/BRCA2-containing complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018055.2 | Intron | NP_001018065.1 | ||||
NM_001242640.1 | Intron | NP_001229569.1 | ||||
NM_024332.3 | Intron | NP_077308.1 | ||||
XM_005274751.4 | Intron | XP_005274808.1 | ||||
XM_017029838.1 | Intron | XP_016885327.1 |
Set Membership: |
HapMap |