Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTCCTCCTGGAGCCACACCCAC[A/G]ATGCCTGGCGGATATCTAAGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602619 MIM: 611749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTBP2 - C-terminal binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083914.2 | 3381 | Intron | NP_001077383.1 | |||
NM_001290214.2 | 3381 | Intron | NP_001277143.1 | |||
NM_001290215.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | NP_001277144.1 | |
NM_001321012.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | NP_001307941.1 | |
NM_001321013.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | NP_001307942.1 | |
NM_001321014.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | NP_001307943.1 | |
NM_001329.3 | 3381 | Intron | NP_001320.1 | |||
NM_022802.2 | 3381 | Silent Mutation | ATC,ATT | I,I 931 | NP_073713.2 | |
XM_005269561.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269618.1 | |
XM_005269564.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269621.1 | |
XM_005269567.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269624.1 | |
XM_005269568.4 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269625.1 | |
XM_005269569.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269626.1 | |
XM_005269571.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269628.1 | |
XM_005269572.3 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_005269629.1 | |
XM_006717642.2 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_006717705.1 | |
XM_011539349.2 | 3381 | Silent Mutation | ATC,ATT | I,I 459 | XP_011537651.1 | |
XM_011539351.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_011537653.1 | |
XM_011539353.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_011537655.1 | |
XM_011539354.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_011537656.1 | |
XM_011539355.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_011537657.1 | |
XM_011539357.1 | 3381 | Silent Mutation | ATC,ATT | I,I 366 | XP_011537659.1 | |
XM_011539358.1 | 3381 | Silent Mutation | ATC,ATT | I,I 366 | XP_011537660.1 | |
XM_017015756.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_016871245.1 | |
XM_017015757.1 | 3381 | Silent Mutation | ATC,ATT | I,I 391 | XP_016871246.1 |
ZRANB1 - zinc finger RANBP2-type containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |