Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGAGAGTGAAGAAAATGATCAGA[C/T]CTTTTCAGAGAAACCAGCATCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616527 | ||||||||||||||||||||
Literature Links: |
CFAP43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP43 - cilia and flagella associated protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFR1 - SWI5 dependent homologous recombination repair protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002759.1 | 296 | Missense Mutation | ACC,ATC | T,I 82 | NP_001002759.1 | |
NM_145247.4 | 296 | Missense Mutation | ACC,ATC | T,I 69 | NP_660290.3 | |
XM_005269521.3 | 296 | Missense Mutation | ACC,ATC | T,I 144 | XP_005269578.1 | |
XM_017015672.1 | 296 | Missense Mutation | ACC,ATC | T,I 69 | XP_016871161.1 |