Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCGCCCCGCACTTCTTCGACAAC[G/T]GGGTCGGCAGCACCAACGGAAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609502 | ||||||||||||||||||||
Literature Links: |
CDHR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDHR1 - cadherin related family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171971.2 | 217 | Missense Mutation | GGG,TGG | G,W 31 | NP_001165442.1 | |
NM_033100.3 | 217 | Missense Mutation | GGG,TGG | G,W 31 | NP_149091.1 | |
XM_011540337.1 | 217 | Missense Mutation | GGG,TGG | G,W 89 | XP_011538639.1 | |
XM_011540338.1 | 217 | Missense Mutation | GGG,TGG | G,W 89 | XP_011538640.1 | |
XM_011540339.1 | 217 | UTR 5 | XP_011538641.1 | |||
XM_011540340.2 | 217 | Missense Mutation | GGG,TGG | G,W 89 | XP_011538642.1 |