Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTTCTTCAATTTCCGTAAGAT[C/T]TTCAAGTTGGGGAGCGAGAAGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616563 | ||||||||||||||||||||
Literature Links: |
SLK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLK - STE20 like kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304743.1 | 600 | Silent Mutation | ATC,ATT | I,I 9 | NP_001291672.1 | |
NM_014720.3 | 600 | Silent Mutation | ATC,ATT | I,I 9 | NP_055535.2 | |
XM_011540401.2 | 600 | Silent Mutation | ATC,ATT | I,I 9 | XP_011538703.1 |