Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTGGTCCTTTTCTGTTAGCTG[C/T]GCTGGAGTTAAGCAAAACAGAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKX - mohawk homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242702.1 | 1099 | Missense Mutation | ACA,GCA | T,A 292 | NP_001229631.1 | |
NM_173576.2 | 1099 | Missense Mutation | ACA,GCA | T,A 292 | NP_775847.2 | |
XM_017016105.1 | 1099 | Missense Mutation | ACA,GCA | T,A 292 | XP_016871594.1 | |
XM_017016106.1 | 1099 | Missense Mutation | ACA,GCA | T,A 292 | XP_016871595.1 | |
XM_017016107.1 | 1099 | Intron | XP_016871596.1 | |||
XM_017016108.1 | 1099 | Intron | XP_016871597.1 |