Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACTTCCCTCATTTGCAGCCAA[A/G]CTTGGGGTGAAGGGCTCCTTTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605677 | ||||||||||||||||||||
Literature Links: |
ACSL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSL5 - acyl-CoA synthetase long-chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016234.3 | 1938 | Silent Mutation | AAA,AAG | K,K 655 | NP_057318.2 | |
NM_203379.1 | 1938 | Silent Mutation | AAA,AAG | K,K 599 | NP_976313.1 | |
NM_203380.1 | 1938 | Silent Mutation | AAA,AAG | K,K 599 | NP_976314.1 | |
XM_017016324.1 | 1938 | Silent Mutation | AAA,AAG | K,K 655 | XP_016871813.1 |
ZDHHC6 - zinc finger DHHC-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303134.1 | 1938 | Intron | NP_001290063.1 | |||
NM_022494.2 | 1938 | Intron | NP_071939.1 | |||
XM_005270051.4 | 1938 | Intron | XP_005270108.1 | |||
XM_006717948.3 | 1938 | Intron | XP_006718011.1 | |||
XM_006717949.3 | 1938 | Intron | XP_006718012.1 | |||
XM_011540091.2 | 1938 | Intron | XP_011538393.1 | |||
XM_011540092.2 | 1938 | Intron | XP_011538394.1 | |||
XM_011540093.1 | 1938 | Intron | XP_011538395.1 | |||
XM_011540094.2 | 1938 | Intron | XP_011538396.1 | |||
XM_011540095.2 | 1938 | Intron | XP_011538397.1 | |||
XM_017016561.1 | 1938 | Intron | XP_016872050.1 | |||
XM_017016562.1 | 1938 | Intron | XP_016872051.1 | |||
XM_017016563.1 | 1938 | Intron | XP_016872052.1 | |||
XM_017016564.1 | 1938 | Intron | XP_016872053.1 | |||
XM_017016565.1 | 1938 | Intron | XP_016872054.1 | |||
XM_017016566.1 | 1938 | Intron | XP_016872055.1 | |||
XM_017016567.1 | 1938 | Intron | XP_016872056.1 | |||
XM_017016568.1 | 1938 | Intron | XP_016872057.1 | |||
XM_017016569.1 | 1938 | Intron | XP_016872058.1 |