Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGCGATATGAGGCAGCATCGA[A/C]AATTTATGGACCGCACGCATTATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ASAH2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASAH2B - N-acylsphingosine amidohydrolase 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079516.3 | 341 | Missense Mutation | AAA,CAA | K,Q 6 | NP_001072984.1 | |
NM_001321957.1 | 341 | Missense Mutation | AAA,CAA | K,Q 6 | NP_001308886.1 | |
NM_001321958.1 | 341 | UTR 5 | NP_001308887.1 | |||
NM_001321959.1 | 341 | UTR 5 | NP_001308888.1 | |||
NM_001321960.1 | 341 | UTR 5 | NP_001308889.1 |
LOC729047 - uncharacterized LOC729047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |