Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGTATGCAAGCCTATAGTCAT[C/T]GAAACTCAGCTCTATGTTATTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGI1 - leucine rich glioma inactivated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308275.1 | 1209 | Intron | NP_001295204.1 | |||
NM_001308276.1 | 1209 | Silent Mutation | ATC,ATT | I,I 243 | NP_001295205.1 | |
NM_005097.3 | 1209 | Silent Mutation | ATC,ATT | I,I 291 | NP_005088.1 | |
XM_017016911.1 | 1209 | Intron | XP_016872400.1 | |||
XM_017016912.1 | 1209 | Intron | XP_016872401.1 |