Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTTGATTAGCTGATTTAAATC[C/T]TCATTCAAAAATGAATCCCCTTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610769 MIM: 608414 | ||||||||||||||||||||
Literature Links: |
NOC3L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOC3L - NOC3 like DNA replication regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022451.10 | 2501 | Silent Mutation | GAA,GAG | E,E 770 | NP_071896.8 | |
XM_017016544.1 | 2501 | Silent Mutation | GAA,GAG | E,E 555 | XP_016872033.1 |
PLCE1 - phospholipase C epsilon 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |