Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACGGCCTTGGCTGCTTGGTCGGA[C/G]GCGCCTACACTTACGGCGGCGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186770 MIM: 612734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLX1 - T-cell leukemia homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195517.1 | 395 | Missense Mutation | CGC,GGC | R,G 53 | NP_001182446.1 | |
NM_005521.3 | 395 | Missense Mutation | CGC,GGC | R,G 53 | NP_005512.1 | |
XM_011539744.2 | 395 | Missense Mutation | CGC,GGC | R,G 53 | XP_011538046.1 | |
XM_011539745.2 | 395 | Missense Mutation | CGC,GGC | R,G 53 | XP_011538047.1 |
TLX1NB - TLX1 neighbor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |