Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTCAGGGAAAGCAAATAAAAAG[A/G]ATACACATGACGGGCCACCAAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC83 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC83 - coiled-coil domain containing 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286159.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | NP_001273088.1 | |
NM_173556.4 | 543 | Missense Mutation | AAT,GAT | N,D 11 | NP_775827.2 | |
XM_011544839.2 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_011543141.1 | |
XM_011544840.2 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_011543142.1 | |
XM_011544841.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_011543143.1 | |
XM_011544842.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_011543144.1 | |
XM_011544843.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_011543145.1 | |
XM_011544844.2 | 543 | Intron | XP_011543146.1 | |||
XM_017017354.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_016872843.1 | |
XM_017017355.1 | 543 | Missense Mutation | AAT,GAT | N,D 11 | XP_016872844.1 |