Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCCGCCACGGTCAGGTTGAGTA[G/T]CCCGCACTTGCGCACACAGTCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603797 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST1 - carbohydrate sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003654.5 | 1229 | Missense Mutation | ATA,CTA | I,L 187 | NP_003645.1 | |
XM_006718356.3 | 1229 | Missense Mutation | ATA,CTA | I,L 187 | XP_006718419.1 | |
XM_017018459.1 | 1229 | Missense Mutation | ATA,CTA | I,L 187 | XP_016873948.1 |