Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCCTGGCTGGAGAGACTGGGC[A/G]TGGGGGTCAATGCACCAGGCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614281 MIM: 602350 MIM: 606011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESAM - endothelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138961.2 | 595 | Missense Mutation | ACG,ATG | T,M 334 | NP_620411.2 | |
XM_005271720.4 | 595 | Missense Mutation | ACG,ATG | T,M 155 | XP_005271777.1 | |
XM_011543060.2 | 595 | Missense Mutation | ACG,ATG | T,M 231 | XP_011541362.1 |
LOC101929340 - uncharacterized LOC101929340 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRGN - neurogranin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSIG2 - V-set and immunoglobulin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |