Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAAGGAAGATGGCACTAAGGA[A/G]AGCCAGCAGGACATAGCGGCCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC37A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC37A2 - solute carrier family 37 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM218 - transmembrane protein 218 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080546.2 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001074015.1 | |
NM_001258238.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245167.1 | |
NM_001258239.1 | 888 | Missense Mutation | CTC,TTC | L,F 121 | NP_001245168.1 | |
NM_001258240.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245169.1 | |
NM_001258241.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245170.1 | |
NM_001258242.1 | 888 | Missense Mutation | CTC,TTC | L,F 121 | NP_001245171.1 | |
NM_001258243.1 | 888 | Missense Mutation | CTC,TTC | L,F 121 | NP_001245172.1 | |
NM_001258244.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245173.1 | |
NM_001258245.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245174.1 | |
NM_001258246.1 | 888 | Missense Mutation | CTC,TTC | L,F 121 | NP_001245175.1 | |
NM_001258247.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | NP_001245176.1 | |
XM_006718779.3 | 888 | Missense Mutation | CTC,TTC | L,F 121 | XP_006718842.1 | |
XM_006718780.3 | 888 | Missense Mutation | CTC,TTC | L,F 121 | XP_006718843.1 | |
XM_006718781.3 | 888 | Missense Mutation | CTC,TTC | L,F 121 | XP_006718844.1 | |
XM_006718783.3 | 888 | Missense Mutation | CTC,TTC | L,F 86 | XP_006718846.1 | |
XM_017017322.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | XP_016872811.1 | |
XM_017017323.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | XP_016872812.1 | |
XM_017017324.1 | 888 | Missense Mutation | CTC,TTC | L,F 86 | XP_016872813.1 |