Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTTACGCAAGGGTGAGGAAGGC[C/T]GCGTTGCTCCCATAGAGGAAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC43A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC43A3 - solute carrier family 43 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278201.1 | 1562 | Silent Mutation | NP_001265130.1 | |||
NM_001278206.1 | 1562 | Silent Mutation | NP_001265135.1 | |||
NM_014096.3 | 1562 | Silent Mutation | NP_054815.2 | |||
NM_017611.2 | 1562 | Silent Mutation | NP_060081.1 | |||
NM_199329.2 | 1562 | Silent Mutation | NP_955361.1 |