Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGACCTGCGCTGTCGCGCGAGG[A/G]GTCTGGGAGATGGGGTGAGTGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGAT2 - diacylglycerol O-acyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253891.1 | 366 | Missense Mutation | GAG,GGG | E,G 36 | NP_001240820.1 | |
NM_032564.4 | 366 | Missense Mutation | GAG,GGG | E,G 36 | NP_115953.2 | |
XM_011545304.2 | 366 | Intron | XP_011543606.1 |
LOC283214 - uncharacterized LOC283214 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |