Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCATCTCAAGGACATTGGATTT[A/G]ATGAATGCCTTGTCCAGATGTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR52N4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR52N4 - olfactory receptor family 52 subfamily N member 4 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005175.3 | 5045 | Missense Mutation | AAT,GAT | N,D 97 | NP_001005175.3 | |
XM_017017711.1 | 5045 | Missense Mutation | AAT,GAT | N,D 97 | XP_016873200.1 | |
XM_017017712.1 | 5045 | Missense Mutation | AAT,GAT | N,D 97 | XP_016873201.1 | |
XM_017017713.1 | 5045 | Missense Mutation | AAT,GAT | N,D 97 | XP_016873202.1 |