Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGACTACCCCTCCCACGCCGTGC[C/T]CTGGAGTGAGGTCCCTCCACCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603940 MIM: 600050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHBP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EHBP1L1 - EH domain binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099409.1 | 1055 | Intron | NP_001092879.1 | |||
XM_005273867.3 | 1055 | Intron | XP_005273924.1 | |||
XM_005273868.3 | 1055 | Intron | XP_005273925.1 | |||
XM_005273869.3 | 1055 | Intron | XP_005273926.1 | |||
XM_005273870.3 | 1055 | Intron | XP_005273927.1 | |||
XM_006718486.3 | 1055 | Intron | XP_006718549.1 | |||
XM_006718487.3 | 1055 | Intron | XP_006718550.1 | |||
XM_011544886.2 | 1055 | Intron | XP_011543188.1 | |||
XM_017017467.1 | 1055 | Intron | XP_016872956.1 | |||
XM_017017468.1 | 1055 | Intron | XP_016872957.1 | |||
XM_017017469.1 | 1055 | Intron | XP_016872958.1 |
KCNK7 - potassium two pore domain channel subfamily K member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005714.1 | 1055 | UTR 3 | NP_005705.1 | |||
NM_033347.1 | 1055 | Intron | NP_203133.1 | |||
NM_033348.1 | 1055 | Missense Mutation | AGC,GGC | S,G 247 | NP_203134.1 | |
NM_033455.1 | 1055 | Missense Mutation | AGC,GGC | S,G 247 | NP_258416.1 |
MAP3K11 - mitogen-activated protein kinase kinase kinase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |