Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTTTCAAAGGCAGGGTTACTC[C/T]ACCCAAGAAAAAATTCTCCCGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3C2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3C2A - phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321378.1 | 5208 | Missense Mutation | AGA,GGA | R,G 1660 | NP_001308307.1 | |
NM_001321380.1 | 5208 | Missense Mutation | AGA,GGA | R,G 1280 | NP_001308309.1 | |
NM_002645.3 | 5208 | Missense Mutation | AGA,GGA | R,G 1660 | NP_002636.2 | |
XM_011520186.2 | 5208 | Missense Mutation | AGA,GGA | R,G 1604 | XP_011518488.1 | |
XM_017017924.1 | 5208 | Missense Mutation | AGA,GGA | R,G 1660 | XP_016873413.1 |