Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGTGTCTGCCAACCTCCCTAAT[C/T]GCACCTTCCACAAACGACAGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TSGA10IP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSGA10IP - testis specific 10 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152762.2 | 1351 | Missense Mutation | CGC,TGC | R,C 374 | NP_689975.2 | |
XM_011544887.2 | 1351 | Missense Mutation | CGC,TGC | R,C 374 | XP_011543189.1 | |
XM_011544888.2 | 1351 | Missense Mutation | CGC,TGC | R,C 238 | XP_011543190.1 |