Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTCCAGGTGGAGGTTTAGCC[A/G]TTGGAACAGTTCTTCTAATTGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELP4 - elongator acetyltransferase complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288725.1 | 294 | Missense Mutation | ATT,GTT | I,V 80 | NP_001275654.1 | |
NM_001288726.1 | 294 | Missense Mutation | ATT,GTT | I,V 80 | NP_001275655.1 | |
NM_019040.4 | 294 | Missense Mutation | ATT,GTT | I,V 80 | NP_061913.3 |