Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAATGGAATCTGGGAAGTAATG[C/G]CAAGGACCCTCGAGGGATGTATCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186790 MIM: 186740 MIM: 603753 | ||||||||||||||||||||
Literature Links: |
CD3D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD3D - CD3d molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD3G - CD3g molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000073.2 | 313 | Missense Mutation | GCC,GGC | A,G 78 | NP_000064.1 | |
XM_005271724.3 | 313 | Missense Mutation | GCC,GGC | A,G 78 | XP_005271781.1 | |
XM_006718941.2 | 313 | Missense Mutation | GCC,GGC | A,G 78 | XP_006719004.1 |
UBE4A - ubiquitination factor E4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |