Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTGTCTTCGATCATGCATTAAT[G/T]AAACATCCACAATTTTTAAAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602286 | ||||||||||||||||||||
Literature Links: |
SC5D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SC5D - sterol-C5-desaturase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024956.2 | 410 | Missense Mutation | ATG,ATT | M,I 63 | NP_001020127.1 | |
NM_006918.4 | 410 | Missense Mutation | ATG,ATT | M,I 63 | NP_008849.2 |