Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTGCCCACACCCATGGCAGAG[C/T]GTTCCCCGGCGGTGATGGTAATGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605390 | ||||||||||||||||||||
Literature Links: |
LPXN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPXN - leupaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143995.2 | 1126 | Silent Mutation | ACA,ACG | T,T 332 | NP_001137467.1 | |
NM_001307951.1 | 1126 | Silent Mutation | ACA,ACG | T,T 307 | NP_001294880.1 | |
NM_004811.2 | 1126 | Silent Mutation | ACA,ACG | T,T 327 | NP_004802.1 | |
XM_006718750.1 | 1126 | Silent Mutation | ACA,ACG | T,T 231 | XP_006718813.1 | |
XM_011545388.2 | 1126 | Silent Mutation | ACA,ACG | T,T 307 | XP_011543690.1 | |
XM_011545390.2 | 1126 | Silent Mutation | ACA,ACG | T,T 307 | XP_011543692.1 | |
XM_011545391.2 | 1126 | Silent Mutation | ACA,ACG | T,T 307 | XP_011543693.1 | |
XM_011545393.2 | 1126 | Intron | XP_011543695.1 | |||
XM_011545394.2 | 1126 | Intron | XP_011543696.1 | |||
XM_017018579.1 | 1126 | Intron | XP_016874068.1 |