Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCAAGCGCTGGCGCAGCTCGT[C/T]GCTGTAGGGGGCCAGATGCGTGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107680 MIM: 107720 MIM: 614776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOA1 - apolipoprotein A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000039.2 | 645 | Missense Mutation | AAC,GAC | N,D 192 | NP_000030.1 | |
NM_001318017.1 | 645 | Missense Mutation | AAC,GAC | N,D 192 | NP_001304946.1 | |
NM_001318018.1 | 645 | Missense Mutation | AAC,GAC | N,D 192 | NP_001304947.1 | |
NM_001318021.1 | 645 | Missense Mutation | AAC,GAC | N,D 83 | NP_001304950.1 |
APOA1-AS - APOA1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOC3 - apolipoprotein C3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIK3 - SIK family kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |